Learn More
LaMotte™ Soil Texture Unit
SDPSCI_ED
Catalog No. S85030 Shop All LaMotte Company Products
Change view
Click to view available options
Product Type:
Reagent Refill
Soil Texture Unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S85030 Soil Texture Unit
S85030R Reagent Refill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S85030 Supplier LaMotte™ Supplier No. 1067
Add to Cart
Add to Cart
Includes
A Study of Soil Science downloadable handbook.
For testing and demonstrating different soil textures and properties.
  • Test soil from school yard or samples brought from home
  • Determines the percentage of sand, silt and clay
  • Overall texture of a soil affects growth in the root zone, which determines the above-ground growth production, and is determined by the fractions of sand, silt, and clay present
Specifications
Class Size 1 Class
Product Type Soil Texture Unit