Learn More
Andwin Scientific SOL BASIN NS PVC 55ML 100/CS
SDP
Catalog No. NC9564797
Encompass_Preferred
The 55 mL pipettor basin is a disposable item that is compatible to use with 8 and 12 channel pipettors. With a capacity of 55 mL and a "V" shaped bottom, this product provides almost complete solution withdrawal. It is constructed with polyvinyl chloride that permits clear viewing of the basins' contents. This product is sold in a case of 100 basins measuring 14.3 x 7 x 2.5 cm (55/8 x 23/4 x 1").
Catalog No. NC9564797 Supplier Andwin Scientific Supplier No. 730001
The 55 mL pipettor basin is a disposable item that is compatible to use with 8 and 12 channel pipettors. With a capacity of 55 mL and a "V" shaped bottom, this product provides almost complete solution withdrawal. It is constructed with polyvinyl chloride that permits clear viewing of the basins' contents. This product is sold in a case of 100 basins measuring 14.3 x 7 x 2.5 cm (55/8 x 23/4 x 1").