Learn More
Andwin Scientific SOL BASIN STRL PS 55ML 80/CSS
SDP
Catalog No. NC9564799
Encompass_Preferred
The 55 mL pipettor basin is a disposable item that is compatible to use with 8 and 12 channel pipettors. With a capacity of 55 mL and a "V" shaped bottom, this product provides almost complete solution withdrawal. It is sterile, is free of RNase, DNase, and pyrogen, and is made of polystrene. This product is sold in a pack of 80 pre-sterilized, individually wrapped basins measuring 14.3 x 7 x 2.5 cm (55/8 x 23/4 x 1").
Catalog No. NC9564799 Supplier Andwin Scientific Supplier No. 730004
The 55 mL pipettor basin is a disposable item that is compatible to use with 8 and 12 channel pipettors. With a capacity of 55 mL and a "V" shaped bottom, this product provides almost complete solution withdrawal. It is sterile, is free of RNase, DNase, and pyrogen, and is made of polystrene. This product is sold in a pack of 80 pre-sterilized, individually wrapped basins measuring 14.3 x 7 x 2.5 cm (55/8 x 23/4 x 1").