Learn More
Quality Biological Inc Solution bottles 250mL, sterile, 24/CS
SDP
Supplier:  Quality Biological Inc P886229782
Encompass_Preferred
CELLTREAT (229782) Solution Bottle, 250mL. Polystyrene bottle with polyethelene cap. Indented grip area. Compatible with bottle top filter units or for solution storage. Sterile (gamma irradiated). 24/CS.
Catalog No. 50-751-7870