Learn More
Sol-Millennium Medical Sol-Care™ Adhesive Foam Dressing
Catalog No. 22025782
Change view
Click to view available options
Dimensions (L x W):
4 x 4 in.
Catalog No. Dimensions (L x W)
22-025-782 4 x 4 in.
1 options
Catalog No. 22-025-782 Supplier Sol-Millennium Medical Supplier No. FMD6110N
Add to Cart
Add to Cart
Foam Dressings made with hydrophilic polyurethane foam for a soft and highly comfortable foam dressing that absorbs exudate and maintains moist wound environment to promote healing.
The product is also made with a polyurethane backing film, skin-friendly acrylic adhesive, and a border to create a seal around the wound. Foam micro-pores with interconnecting channels enable efficient and reliable vertical fluid absorption, tailored to wound care needs. The uniformly distributed pores effectively manage high and viscous exudate volumes, providing consistent absorption across various wound conditions. The waterproof outer layer acts as a barrier against bacterial and viral penetration, providing confidence in infection prevention.
  • Breathable
  • Waterproof
  • Highly Absorbent
  • Cushioned for Skin
  • Change Frequency: Up to 7 Days
  • Indications: Sol-Care™ Foam Dressing is designed for moderate to heavy exuding wound management by secondary intention on shallow, granulating wounds, chronic and acute exudative wounds, full and partial thickness wounds such as pressure injuries, diabetic foot ulcers, venous and arterial leg ulcers, surgical wounds and donor sites.
Specifications
Product Type Adhesive Bandage
Quantity 200/Cs.
Dimensions (L x W) 4 x 4 in.
Latex Free Yes