Learn More
Sol-Millennium Medical Sol-Care™ Alginate Dressing, Rope
Catalog No. 22025780
Change view
Click to view available options
Dimensions (L x W):
12 x 0.75 in.
Catalog No. Dimensions (L x W)
22-025-780 12 x 0.75 in.
1 options
Catalog No. 22-025-780 Supplier Sol-Millennium Medical Supplier No. FAD1210N
Add to Cart
Add to Cart
Designed for moderately to high exuding wounds, such as diabetic foot ulcers, pressure injuries, venous and arterial leg ulcers, superficial burns, donor sites, post-operative wounds and traumatic wounds.
  • Biodegradable
  • High Absorbent
  • Can be Cut
  • Change Frequency: Up to 7 Days
Specifications
Product Type Gauze Bandage
Quantity 400/Cs.
Dimensions (L x W) 12 x 0.75 in.
Latex Free Yes