Learn More
Sol-Millennium Medical Sol-Care™ Alginate Dressing
Catalog No. 22025779
Change view
Click to view available options
Dimensions (L x W):
4 x 4 in.
Catalog No. Dimensions (L x W)
22-025-779 4 x 4 in.
1 options
Catalog No. 22-025-779 Supplier Sol-Millennium Medical Supplier No. FAD1120N
Add to Cart
Add to Cart
Designed for moderately to high exuding wounds, such as diabetic foot ulcers, pressure injuries, venous and arterial leg ulcers, superficial burns, donor sites, post-operative wounds and traumatic wounds.
  • Biodegradable
  • High Absorbent
  • Can be Cut
  • Change Frequency: Up to 7 Days
Specifications
Product Type Gauze Bandage
Quantity 800/Cs.
Dimensions (L x W) 4 x 4 in.
Latex Free Yes