Learn More
Sol-Millennium Medical Sol-Care™ Hydrocolloid Dressing, Thin
Catalog No. 22025785
Change view
Click to view available options
Dimensions (L x W):
4 x 4 in.
Catalog No. Dimensions (L x W)
22-025-785 4 x 4 in.
1 options
Catalog No. 22-025-785 Supplier Sol-Millennium Medical Supplier No. HCD2120N
Add to Cart
Add to Cart
Hydrocolloid Dressings create and maintain a moist wound environment by absorbing wound exudate to form a soft gel without swelling.
The polyurethane film allows excess moisture to transpire through it, thus enhancing the ability of the dressing to manage exudate absorption. The smooth outer surface guards against shearing and friction, protecting the surrounding skin. The durable and body-conforming design with tapered edges helps to prevent shifting or rolling, ensuring long-term wear and comfort. The waterproof barrier prevents external contamination and maintains the dressing's integrity.
  • Waterproof
  • Translucent
  • Self-Adhesive
  • Change Frequency: Up to 7 Days
  • Indications: Sol-Care™ Hydrocolloid Dressing is designed for a wide range of non-moderate exuding chronic and acute wounds, such as pressure injuries, leg ulcers, donor sites, postoperative wounds, and skin abrasions.
Specifications
Product Type Hydrocolloid Dressing
Quantity 600/Cs.
Dimensions (L x W) 4 x 4 in.
Latex Free Yes