Learn More
Sol-Millennium Medical Sol-Care™ IV Transparent Film Dressing, Border
Catalog No. 22025919
Change view
Click to view available options
Dimensions (L x W):
2.25 x 2 in.
Catalog No. Dimensions (L x W)
22-025-919 2.25 x 2 in.
1 options
Catalog No. 22-025-919 Supplier Sol-Millennium Medical Supplier No. TFD7110N
Add to Cart
Add to Cart
It can be used to cover and protect catheter sites, as a secondary dressing as a protective cover over at-risk skin to secure devices to the skin, or to provide a moist wound environment to facilitate autolytic debridement.
The product features a mild adhesive that provides reliable fixation while comfortably conforming to body contours, enabling a soft and pain-free dressing application. They can be cut to any size or shape of the wound, offering flexibility in various clinical applications. The transparent design allows for easy, non-invasive monitoring of wounds and I.V. sites without the need to remove the dressing. The transparent dressings are breathable and waterproof, they have a sterile barrier that is impervious to liquids, bacteria, and viruses.
  • Waterproof
  • Transparent
  • Versatile
  • Change Frequency: Up to 7 Days
  • Indications: Sol-Care™ Transparent Film Dressing is designed to cover, fix and protect IV sites and wounds. It is also suitable as a secondary dressing, a protective barrier for at-risk skin, or securely fastening devices to the skin.
Specifications
Product Type Transparent Film Dressing
Quantity 2000/Cs.
Dimensions (L x W) 2.25 x 2 in.
Color White
Latex Free Yes