Learn More
Sol-Millennium Medical Sol-Care™ Safety Blood Collection Needle 3/4 in. with 12 in. Tubing and Luer Adapter
Catalog No. 22025859
Change view
Click to view available options
Needle Gauge:
21 G
25 G
Color:
Green
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
22-025-859 25 G Orange
22-025-855 21 G Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-025-859 Supplier Sol-Millennium Medical Supplier No. SBCN2512
Add to Cart
Add to Cart
Blood Collection Needle is a safety-engineered device with a sliding safety shield that clicks when activated, alerting you that the needle is safely locked.
  • Transparent design that allows visualization of the blood flashback and flow
  • Single-handed activation option that provides more flexibility to care for the patient and venipuncture site
  • Sliding safety shield pushes over the needle and locks with an audible click, protecting against needle-stick injury
Specifications
Product Type Blood Collection Set
Color Orange
Material Plastic, Stainless Steel
Needle Gauge 25 G
Sterility Sterile
Includes 12 in. Tubing
Latex Free Yes