Learn More
ICU Medical Sol-Care™ Safety Blood Collection Needle Infusion Set
Catalog No. 22024016
Encompass
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22-024-016 21 G
22-024-017 23 G
22-024-018 25 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-024-016 Supplier ICU Medical Supplier No. 213412SI
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safety butterfly needle designed to be used for blood collection.
  • Simple safety mechanism that should be activated immediately after blood withdrawal is complete
  • Reduces the risk of needlestick injury
  • Thumb rest to ease handling during safety activation
  • Flexible wings to secure device to patient's skin
  • Thin walled needle is designed to minimize patient discomfort
  • Available in 21, 23 or 25 gauge
  • Transparent housing and tubing for clear visualization of blood flow
  • DEHP-free, flexible, soft, and kink resistant tubing
Specifications
Product Type Needle Infusion Set
Color Green
Material PVC Tubing
Needle Gauge 21 G
Quantity 50 Pk., 4 Pk./Cs.