Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sol-Millennium Medical Sol-Care™ Safety Pen Needle

Catalog No. 22025914
Change view
Click to view available options
Needle Gauge:
30 G
31 G
32 G
Length (Metric) Needle:
4 mm
5 mm
6 mm
8 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (Metric) Needle
22-025-914 32 G 4 mm
22-025-910 30 G 5 mm
22-025-911 30 G 8 mm
22-025-913 31 G 5 mm
22-025-912 31 G 6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-025-914 Supplier Sol-Millennium Medical Supplier No. SPNS32532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intended for use with pen injectors for subcutaneous delivery of insulin.

  • Shields lock automatically after injection, helping to prevent accidental injury during disposal
  • Needle front pre-covered for safety
  • Red safety visual to indicate once safety mechanism has been activated
  • Fits most popular pens on the market
  • Wider surface area helps minimize pressure during injection

Specifications

Material Polypropylene, Stainless Steel
Needle Gauge 32 G
Latex Free Yes
Product Type Pen Needle
Quantity 800/Cs.
Length (Metric) Needle 4 mm
For Use With (Equipment) Insulin Pens
Connection Type Female Luer
Graduated No
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.