Learn More
Sol-Millennium Medical Sol-Care™ Safety Winged Blood Collection Needle with Pre-attached Holder 3/4 in. and with 7 in. Tubing
Description
- Transparent design that allows visualization of the blood flashback and flow
- Single-handed activation option that provides more flexibility to care for the patient and venipuncture site
- Sliding safety shield pushes over the needle and locks with an audible click, protecting against needle-stick injury
Specifications
Specifications
| Product Type | Blood Collection Set |
| Color | Orange |
| Material | Plastic, Stainless Steel |
| Needle Gauge | 25 G |
| Sterility | Sterile |
| Latex Free | Yes |