Learn More
Sol-Millennium Medical Sol-Care™ Silicone Foam Dressing, Border
Description
The extra soft silicone wound contact layer forms a seal between the dressing and the wound, ensuring that would exudate is absorbed into the dressing while reducing trauma and pain upon removal. The foam layer cushions, reduces pressure, friction, and other shear forces, and improves patient comfort while lowering risk of further injury. The silicone adhesive forms a secure bond that is gentle on the wound and surrounding skin, allowing for safe removal. The super-absorbent fiber layer vertically captures and retains exudate, maintaining a clean, optimal healing environment. The breathable construction promotes moisture vapor transmission, supporting a healthy wound environment and preventing excess moisture buildup around the peri-wound skin.
- Cushioned for skin
- Gentle silicone Self-adhesion
- Change Frequency: Up to 7 days
- Indications: The Sol-Care™ Silicone Foam Dressing is designed for a wide range of exuding wounds such as pressure injuries, venous and arterial leg ulcers, diabetic foot ulcers, and traumatic and surgical wounds.
Specifications
Specifications
| Product Type | Adhesive Bandage |
| Quantity | 400/Cs. |
| Dimensions (L x W) | 4 x 4 in. |
| Color | Beige |
| Latex Free | Yes |