Learn More
Sol-Millennium Medical Sol-Care™ Silicone Foam Dressing
Catalog No. 22025873
Change view
Click to view available options
Dimensions (L x W):
4 x 4 in.
Catalog No. Dimensions (L x W)
22-025-873 4 x 4 in.
1 options
Catalog No. 22-025-873 Supplier Sol-Millennium Medical Supplier No. SFD2110N
Add to Cart
Add to Cart
Silicone Foam Dressings are made from a foam layer, a super-absorbent fiber layer, and a nonwoven layer, a polyurethane backing film, and a perforated silicone wound contact layer.
The extra soft silicone wound contact layer forms a seal between the dressing and the wound, ensuring that would exudate is absorbed into the dressing while reducing trauma and pain upon removal. The foam layer cushions, reduces pressure, friction, and other shear forces, and improves patient comfort while lowering risk of further injury. The silicone adhesive forms a secure bond that is gentle on the wound and surrounding skin, allowing for safe removal. The super-absorbent fiber layer vertically captures and retains exudate, maintaining a clean, optimal healing environment. The breathable construction promotes moisture vapor transmission, supporting a healthy wound environment and preventing excess moisture buildup around the peri-wound skin.
  • Cushioned for skin
  • Gentle silicone Self-adhesion
  • Change Frequency: Up to 7 days
  • Indications: The Sol-Care™ Silicone Foam Dressing is designed for a wide range of exuding wounds such as pressure injuries, venous and arterial leg ulcers, diabetic foot ulcers, and traumatic and surgical wounds.
Specifications
Product Type Adhesive Bandage
Quantity 400/Cs.
Dimensions (L x W) 4 x 4 in.
Color Beige
Latex Free Yes