Learn More
Sol-Millennium Medical Sol-Care™ Super Absorbent Dressing
Catalog No. 22025853
Change view
Click to view available options
Dimensions (L x W):
4 x 4 in.
Catalog No. Dimensions (L x W)
22-025-853 4 x 4 in.
1 options
Catalog No. 22-025-853 Supplier Sol-Millennium Medical Supplier No. SAP1110N
Add to Cart
Add to Cart
Super Absorbent Dressings allow for maximum absorption as they contain SAP (Super Absorbent Polymer).
The multi-layer system efficiently absorbs and locks in exudate, preventing leakage and supporting better wound care. The high absorbency maintains the ideal moisture balance, protecting the wound edges from maceration. The SMS non-woven material enhances patient comfort and offers excellent transpiration. The water-repellent and anti-bacterial outer layer protects against fluid strike-through and provides an additional barrier against bacteria.
  • Maximum Absorbency
  • Locks in Drainage
  • Change Frequency: Up to 7 Days
  • Indications: Sol-Care™ Super Absorbent Dressing is designed for acute and chronic wounds with moderate to high exudate levels, such as diabetic foot ulcers, pressure injuries, venous and arterial leg ulcers, first and second-degree burns, postoperative wounds, and traumatic wounds.
Specifications
Product Type Gauze Bandage
Sterility Sterile
Quantity 800/Cs.
Dimensions (L x W) 4 x 4 in.
Latex Free Yes