Learn More
Med Vet International Diabetes Management Test Control Assure Prism Blood Glucose Test 2 Levels, 530006, 1/Box
Catalog No. 503045383
Encompass_Preferred
Assure Prism multi control solution ensures that your meter and test strips are functioning correctly The solution is compatible with Assure Prism Blood glucose meter and test strips The high accuracy of the blood glucose meter control solution ensures that the results given by the meter are precise The solution helps make sure you perform tests the right way for greater accuracy Assure Prism control solution includes one bottle each of level one and level two solution
Catalog No. 50-304-5383 Supplier Med Vet International Supplier No. 971974BX
Assure Prism multi control solution ensures that your meter and test strips are functioning correctly The solution is compatible with Assure Prism Blood glucose meter and test strips The high accuracy of the blood glucose meter control solution ensures that the results given by the meter are precise The solution helps make sure you perform tests the right way for greater accuracy Assure Prism control solution includes one bottle each of level one and level two solution