Learn More
Med Vet International 3M Bitter Fit Test Solution, Ft-32, 6/Case
Catalog No. 502831117
Encompass_Preferred
This qualitative fit test solution helps you make sure your respirator functions right Its bitter taste makes it quick and easy to discern It's compatible for use with any particular respirator or gas and vapor respirator This 3M™ fit test solution is easy to use - just spray it on the hood of the respirator that's being tested Meets OSHA standards to make sure your facility is safe It's ideal for use with the 3M™ Qualitative Fit Test Apparatus FT-30
Catalog No. 50-283-1117 Supplier Med Vet International Supplier No. 352434CS
This qualitative fit test solution helps you make sure your respirator functions right Its bitter taste makes it quick and easy to discern It's compatible for use with any particular respirator or gas and vapor respirator This 3M™ fit test solution is easy to use - just spray it on the hood of the respirator that's being tested Meets OSHA standards to make sure your facility is safe It's ideal for use with the 3M™ Qualitative Fit Test Apparatus FT-30