Learn More
Med Vet International 3M Sweet Fit Test Solution, Ft-12, 6/Case
Catalog No. 502830035
Encompass_Preferred
The 3M™ sweet fit test solution works with the 3M™ FT-10 Fit Test Apparatus to help respirators meet OSHA standards Its design allows organizations to meet regulatory fit-testing requirements for respirators The respirator test solution works with a nebulizer and collar to test the fit of dust masks and respirators It's also made for the 3M™ Training and Fit-Testing Case FT-20This 3M™ qualitative fit test uses water and sodium saccharine to produce a sweet-tasting aerosol
Catalog No. 50-283-0035 Supplier Med Vet International Supplier No. 235410CS
The 3M™ sweet fit test solution works with the 3M™ FT-10 Fit Test Apparatus to help respirators meet OSHA standards Its design allows organizations to meet regulatory fit-testing requirements for respirators The respirator test solution works with a nebulizer and collar to test the fit of dust masks and respirators It's also made for the 3M™ Training and Fit-Testing Case FT-20This 3M™ qualitative fit test uses water and sodium saccharine to produce a sweet-tasting aerosol