Learn More
Sol-Millennium Medical Sol-Guard™ Safety Pull-button Blood Collection Set 3/4 7 in.
Catalog No. 22025849
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
Color:
Blue
Green
Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
22-025-849 25 G Orange
22-025-847 21 G Green
22-025-848 23 G Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-025-849 Supplier Sol-Millennium Medical Supplier No. PBLA2507
Add to Cart
Add to Cart
Blood Collection Sets provide improved comfort, quicker procedure times and single-handed in-vein safety activation to instantly retract the needle after use, helping prevent needle-stick injury and bloodborne pathogen exposure.
  • Flashback for instant and clear confirmation of proper needle placement
  • Low angled tri-bevel tip designed to improve penetration, while minimizing patient discomfort
  • Spring mechanism for split second in-vein needle retraction
  • Large oversized arrow to easily identify the activation pull button
  • 30% wider wings for improved handling during insertion and removal
Specifications
Product Type Blood Collection Set
Color Orange
Material Plastic, Stainless Steel
Needle Gauge 25 G
Sterility Sterile
Latex Free Yes