Learn More
Sol-Millennium Medical Sol-M™ 1 mL Luer Lock Syringe without Needle Tray (PP)
Catalog No. 22025760
Change view
Click to view available options
Volume (Metric):
1 mL
Catalog No. Volume (Metric)
22-025-760 1 mL
1 options
Catalog No. 22-025-760 Supplier Sol-Millennium Medical Supplier No. 180001T
Add to Cart
Add to Cart
1 mL syringe designed for accurate liquid measurement and controlled dispensing.
  • Made from polypropylene (PP) for durability and reliable performance.
Specifications
Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge No Needle Included
Latex Free Yes
Product Type Standard Syringe
Volume (Metric) 1 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Luer Lock
Graduated Yes
Sterility Sterile