Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICU Medical SOL-M™ Blood Collection Tube Holder

Catalog No. 22024014
Encompass
Change view
Click to view available options
Size:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-024-014 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-024-014 Supplier ICU Medical Supplier No. 110201050001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for use with SOL-CARE™ Safety Blood Collection Needle with Multi-Sample Luer.

  • Translucent
  • For blood collection
  • For use with SOL-CARE™ Safety Blood Collection Needle with Multi-Sample Luer

Specifications

Color Translucent
Length (English) 2 in.
Material Polypropylene
For Use With (Application) Blood Draw
Length (Metric) 5 cm
Size Standard
Product Type Tube Holder
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.