Learn More
Sol-Millennium Medical Sol-M™ Eccentric Tip Syringe without Needle
Catalog No. 22025837
Change view
Click to view available options
Volume (Metric):
10 mL
20 mL
60 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-025-837 60 mL
22-025-824 10 mL
22-025-829 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-025-837 Supplier Sol-Millennium Medical Supplier No. P180060ET
Add to Cart
Add to Cart
The 3 piece syringes accurately measure liquid medications and are available in a wide range of sizes.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger from the barrel
  • Large finger grips allow for easy, controlled injections
  • Available in Luer Lock, Luer Slip Tip, and Catheter Tip options
Specifications
Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge No Needle Included
Latex Free Yes
Product Type Standard Syringe
Disposable Single-use
Volume (Metric) 60 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Eccentric
Graduated Yes
Sterility Sterile