Learn More
Sol-Millennium Medical Sol-M™ Lancing Device
Catalog No. 22025795
Change view
Click to view available options
For Use With (Equipment):
Lancet
Catalog No. For Use With (Equipment)
22-025-795 Lancet
1 options
Catalog No. 22-025-795 Supplier Sol-Millennium Medical Supplier No. LAD001
Add to Cart
Add to Cart
The lancing device is used for capillary blood sampling required in daily diabetes management programs.
  • Offers 6 settings for depth puncture
  • Red visual indicator confirms when lancing device is ready for use
  • Works with most general lancets
Specifications
For Use With (Application) Diabetes Management
For Use With (Equipment) Lancet
Product Type Fingerstick Incision Device