Learn More
Sol-Millennium Medical Sol-M™ Level 2 Isolation Gowns
Catalog No. 22025789
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-025-789 X-Large
22-025-787 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-025-789 Supplier Sol-Millennium Medical Supplier No. IG004XL
Add to Cart
Add to Cart
Protective Level 2 isolation gowns designed to help support barrier protection in healthcare and clinical environments.
  • AAMI Level 2 isolation gown made from a premium 23 GSM SMS material that is both breathable and fluid-resistant
  • Over-the-head construction makes donning quick and convenient
  • Neck seam is designed for easy tear-away that makes doffing clean and safe for the wearer
  • Elastic Wrist Cuff
Specifications
Size X-Large
Product Type Isolation Gown
Color White
Disposable Limited-use
Latex Free Yes
No. per Pack 100/Cs.