Learn More
Sol-Millennium Medical Sol-M™ Level 3 Isolation Gowns
Catalog No. 22025792
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-025-792 X-Large
22-025-791 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-025-792 Supplier Sol-Millennium Medical Supplier No. IG433XL2133
Add to Cart
Add to Cart
Level 3 Isolation Gowns that are highly fluid-resistant.
  • Isolation gown constructed from poly-coated 45 GSM SPP that is highly fluid-resistant
  • Hook and loop neck closure for easy donning and doffing with durable binding
  • Premium knit cuffs and wide belt ties at the waist
Specifications
Size X-Large
Product Type General Purpose Gown
Color Blue
Disposable Limited-use
Cuff Style Elastic
Latex Free Yes
No. per Pack 50/Cs.