Learn More
Sol-Millennium Medical Sol-M™ Level 4 Chemotherapy Gown
Catalog No. 22025790
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-025-790 X-Large
22-025-788 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-025-790 Supplier Sol-Millennium Medical Supplier No. IG0050
Add to Cart
Add to Cart
AAMI Level 4 isolation gowns that have been tested against chemotherapy drugs for use (ASTM D6978).
  • Non-sterile isolation gowns with hook and loop neck closure, knit cuffs, and full back covering.
Specifications
Size X-Large
Product Type Chemotherapy Gown
Color Blue
Disposable Limited-use
Sterility Non-sterile
Latex Free Yes
No. per Pack 50/Cs.