Learn More
Sol-Millennium Medical Sol-M™ Luer Adapter 20 G
Description
- An ideal accessory for use with blood collection sets that feature a holder for evacuated tubes
- Blue cap to differentiate from other blood collection devices for quick identification to reduce selection errors
- Designed to ensure secure connections and fitted with rubber sleeve to ensure no blood spills or exposure
Specifications
Specifications
| Latex Free | Yes |
| Product Type | Luer Adapter |
| For Use With (Equipment) | Removable Needle Syringe |
| Sterility | Non-sterile |