Learn More
Sol-Millennium Medical Sol-M™ Luer Lock Syringe with Exchangeable Needle 21 G 1 1/2 in.
Description
- High-transparency barrel to easily visualize contents
- Silicone lubrication ensures smooth plunger movement
- Plunger backstop that prevents accidental withdrawal of plunger from the barrel
- Large finger grips allow for easy, controlled injections
- Available in Luer Lock, Luer Slip Tip, and Catheter Tip options
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Stainless Steel, Polypropylene (Barrel) |
| Needle Gauge | 21 G |
| Latex Free | Yes |
| Product Type | Syringe with Exchangeable Needle |
| Volume (Metric) | 5 mL |
| Connection Type | Luer Lock |
| Graduated | Yes |
| Sterility | Sterile |