Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sol-Millennium Medical Sol-M™ Luer Lock Syringe Sterile Convenience Tray

Catalog No. 22025840
Change view
Click to view available options
Volume (Metric):
3 mL
5 mL
10 mL
20 mL
30 mL
60 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-025-840 60 mL
22-025-820 3 mL
22-025-823 5 mL
22-025-827 10 mL
22-025-832 20 mL
22-025-836 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-025-840 Supplier Sol-Millennium Medical Supplier No. P180060TU
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The syringe sterile convenience trays are available in a wide variety of both standard and safety syringes. The trays are intended to be used in pharmacies to aid with batch-filling.

  • Variety of sizes from 1 to 60 mL
  • Clear, bold graduations for accurate filling and dosing
  • Sterile trays for convenient filling and use
  • Available with safety-engineered syringes

Specifications

Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge No Needle Included
Latex Free Yes
Product Type Standard Syringe
Disposable Single-use
Volume (Metric) 60 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Luer Lock
Graduated Yes
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.