Learn More
Sol-Millennium Medical Sol-M™ Oral Syringe Clear with Tip Cap (O-ring, Bulk, Non-sterile)
Description
- 3-piece syringe with O-ring
- Large variety of sizes and colors
- Clear graduation markings for accurate medication delivery
- Not made with natural rubber latex
- Special tip design will not accept Luer connectors, preventing medication errors and the accidental intravenous injections of oral liquids
- Optional amber barrel designed for light-sensitive drugs, in compliance with United States Pharmacopeia (USP) requirements
- Packaged in a PE bag
Specifications
Specifications
| Color | Amber |
| For Use With (Application) | Oral Medication Administration |
| Latex Free | Yes |
| Product Type | Oral Syringe |
| Disposable | Single-use |
| Volume (Metric) | 10 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.