Learn More
Sol-Millennium Medical Sol-M ™ Shoe Cover, 35 GSM
Description
- Made from durable SMS material that comes in both medium and heavy weights
- Anti-skid bands provide traction on the bottom of the covers
- Elasticated and intended to be worn over shoes to prevent the spread of contaminants
Specifications
Specifications
| Size | 2X-Large |
| Material | SMS |
| Latex Free | Yes |
| Color | Blue |
| Disposable | Limited-use |
| Packaging Quantity | 300/Cs. |
| Product Type | Shoe Covers |