Learn More
Sol-Millennium Medical Sol-M™ Slip Tip Sterile Syringe without Needle
Catalog No. 22025834
Change view
Click to view available options
Volume (Metric):
30 mL
Catalog No. Volume (Metric)
22-025-834 30 mL
1 options
Catalog No. 22-025-834 Supplier Sol-Millennium Medical Supplier No. P180030ST
Add to Cart
Add to Cart
The 3-piece bulk, non-sterile syringes accurately measure liquid medications and are available in a wide range of sizes.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger from the barrel
  • Large finger grips allow for easy, controlled injections
  • Available in Luer Lock, Luer Slip Tip, and Catheter Tip options
Specifications
Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge No Needle Included
Latex Free Yes
Product Type Standard Syringe
Disposable Single-use
Volume (Metric) 30 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Luer Slip
Graduated Yes
Sterility Sterile