Learn More
Sol-Millennium Medical Sol-M™ Slip Tip Syringe without Needle (Bulk, Non-sterile)
Catalog No. 22025839
Change view
Click to view available options
Volume (Metric):
3 mL
5 mL
10 mL
20 mL
30 mL
60 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-025-839 60 mL
22-025-819 3 mL
22-025-822 5 mL
22-025-826 10 mL
22-025-831 20 mL
22-025-835 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-025-839 Supplier Sol-Millennium Medical Supplier No. P180060STNSB
Add to Cart
Add to Cart
The 3-piece bulk, non-sterile syringes accurately measure liquid medications and are available in a wide range of sizes.
  • High-transparency barrel to easily visualize contents
  • Silicone lubrication ensures smooth plunger movement
  • Plunger backstop that prevents accidental withdrawal of plunger for the barrel
  • Large finger grips allow for easy, controlled injections
  • Available in Luer Lock, Luer Slip Tip, and Catheter Tip options
  • Packaged in a PE bag
Specifications
Autoclavable Not Autoclavable
Material Polypropylene
Needle Gauge No Needle Included
Latex Free Yes
Product Type Standard Syringe
Disposable Single-use
Volume (Metric) 60 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Luer Slip
Graduated Yes
Sterility Non-sterile