Learn More
Med Vet International 3M Bitter SensitIVity Solution, Bitter, Ft-31, Each
Catalog No. 502831714
Encompass_Preferred
The bitter sensitivity solution helps in checking that there's a safe respiratory seal It's used with the 3M™ FT-30 Fit Test Apparatus to ensure the respirator meets OSHA standards This solution uses denatonium benzoate to produce a bitter taste It allows organizations to meet regulatory fit-testing requirements for respirators This 3M™ sensitivity solution works with a nebulizer and collar to test the fit of dust masks and respirators
Catalog No. 50-283-1714 Supplier Med Vet International Supplier No. 352740EA
The bitter sensitivity solution helps in checking that there's a safe respiratory seal It's used with the 3M™ FT-30 Fit Test Apparatus to ensure the respirator meets OSHA standards This solution uses denatonium benzoate to produce a bitter taste It allows organizations to meet regulatory fit-testing requirements for respirators This 3M™ sensitivity solution works with a nebulizer and collar to test the fit of dust masks and respirators