Learn More
Med Vet International 3M SensitIVity Solution, Saccharin, Ft-11, Each
Catalog No. 502830034
Encompass_Preferred
The 3M™ fit-testing solution ensures the proper fit of face to respirator seal It has the sweet taste of saccharin that if detected by the user identifies an improper fit The sweet 3M™ sensitivity solution is acceptable for particulate, gas or vapor respirators that include a pre-filter It meets or exceeds OSHA safety standards for fit for proper fit of respirators The sensitivity solution fits 3M™ qualitative fit-test apparatus FT-10 and 3M™ training and fit-testing case FT-20
Catalog No. 50-283-0034 Supplier Med Vet International Supplier No. 235409EA
The 3M™ fit-testing solution ensures the proper fit of face to respirator seal It has the sweet taste of saccharin that if detected by the user identifies an improper fit The sweet 3M™ sensitivity solution is acceptable for particulate, gas or vapor respirators that include a pre-filter It meets or exceeds OSHA safety standards for fit for proper fit of respirators The sensitivity solution fits 3M™ qualitative fit-test apparatus FT-10 and 3M™ training and fit-testing case FT-20