Learn More
Med Vet International ReadyPrep Skin Prep Solution, 16 oz. Bottle, 10% Strength, Povidone-Iodine, NonSterile, MDS093906, 24/Case
Catalog No. 503044539
Encompass_Preferred
Effective for skin and mucous membrane preparation prior to surgical procedures Also effective as a first aid antiseptic
Catalog No. 50-304-4539 Supplier Med Vet International Supplier No. 353348CS
Effective for skin and mucous membrane preparation prior to surgical procedures Also effective as a first aid antiseptic