Learn More
Hampton Research SOL & STABILITY #11 (A11)
SDP
Catalog No. 50562965
Encompass
HR2-072-11/Solubility & Stability #11 (A11), 1.0 ml - 500 mM L-Argininamide dihydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-562-965 Supplier Hampton Research Supplier No. HR207211
HR2-072-11/Solubility & Stability #11 (A11), 1.0 ml - 500 mM L-Argininamide dihydrochloride. Please note items from this supplier are non-returnable.