Learn More
Hampton Research SOL & STABILITY #39 (D3)
SDP
Catalog No. 50562993
Encompass
HR2-072-39/Solubility & Stability #39 (D03), 1.0 ml - 5% w/v Benzamidine hydrochloride. Please note items from this supplier are non-returnable.
Catalog No. 50-562-993 Supplier Hampton Research Supplier No. HR207239
HR2-072-39/Solubility & Stability #39 (D03), 1.0 ml - 5% w/v Benzamidine hydrochloride. Please note items from this supplier are non-returnable.