Learn More
Hampton Research SOL & STABILITY #42 (D6)
SDP
Catalog No. 50562996
Encompass
HR2-072-42/Solubility & Stability #42 (D06), 1.0 ml - 100 mM Cadmium chloride hydrate, 100 mM Cobalt(II) chloride hexahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-562-996 Supplier Hampton Research Supplier No. HR207242
HR2-072-42/Solubility & Stability #42 (D06), 1.0 ml - 100 mM Cadmium chloride hydrate, 100 mM Cobalt(II) chloride hexahydrate. Please note items from this supplier are non-returnable.