Learn More
Hampton Research SOL & STABILITY #45 (D9)
SDP
Catalog No. 50562999
Encompass
HR2-072-45/Solubility & Stability #45 (D09), 1.0 ml - 1,000 mM Non Detergent Sulfobetaine 211 (NDSB-211). Please note items from this supplier are non-returnable.
Catalog No. 50-562-999 Supplier Hampton Research Supplier No. HR207245
HR2-072-45/Solubility & Stability #45 (D09), 1.0 ml - 1,000 mM Non Detergent Sulfobetaine 211 (NDSB-211). Please note items from this supplier are non-returnable.