Learn More
Hampton Research SOL & STABILITY #47B (D11)
SDP
Catalog No. 50563002
Encompass
HR2-072-47B/Solubility & Stability #47B (D11), 1.0 ml - 800 mM Non Detergent Sulfobetaine 256 (NDSB-256). Please note items from this supplier are non-returnable.
Catalog No. 50-563-002 Supplier Hampton Research Supplier No. HR207247B
HR2-072-47B/Solubility & Stability #47B (D11), 1.0 ml - 800 mM Non Detergent Sulfobetaine 256 (NDSB-256). Please note items from this supplier are non-returnable.