Learn More
Hampton Research SOL & STABILITY #49 (E1)
SDP
Catalog No. 50563004
Encompass
HR2-072-49/Solubility & Stability #49 (E01), 1.0 ml - 500 mM Acetamide. Please note items from this supplier are non-returnable.
Catalog No. 50-563-004 Supplier Hampton Research Supplier No. HR207249
HR2-072-49/Solubility & Stability #49 (E01), 1.0 ml - 500 mM Acetamide. Please note items from this supplier are non-returnable.