Learn More
Hampton Research SOL & STABILITY #71 (F11)
SDP
Catalog No. 50563025
Encompass
HR2-072-71/Solubility & Stability #71 (F11), 1.0 ml - 250 mM Sodium p-toluenesulfonate. Please note items from this supplier are non-returnable.
Catalog No. 50-563-025 Supplier Hampton Research Supplier No. HR207271
HR2-072-71/Solubility & Stability #71 (F11), 1.0 ml - 250 mM Sodium p-toluenesulfonate. Please note items from this supplier are non-returnable.