Learn More
Hampton Research SOL & STABILITY #74 (G2)
SDP
Catalog No. 50563028
Encompass
HR2-072-74/Solubility & Stability #74 (G02), 1.0 ml - 700 mM Sodium phosphate monobasic monohydrate, 1,300 mM Potassium phosphate dibasic. Please note items from this supplier are non-returnable.
Catalog No. 50-563-028 Supplier Hampton Research Supplier No. HR207274
HR2-072-74/Solubility & Stability #74 (G02), 1.0 ml - 700 mM Sodium phosphate monobasic monohydrate, 1,300 mM Potassium phosphate dibasic. Please note items from this supplier are non-returnable.