Learn More
Hampton Research SOL & STABILITY #78 (G6)
SDP
Catalog No. 50563032
Encompass
HR2-072-78/Solubility & Stability #78 (G06), 1.0 ml - 40% v/v Glycerol, 400 mM Lithium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-563-032 Supplier Hampton Research Supplier No. HR207278
HR2-072-78/Solubility & Stability #78 (G06), 1.0 ml - 40% v/v Glycerol, 400 mM Lithium chloride. Please note items from this supplier are non-returnable.