Learn More
Sol-Millennium Medical Sol-Vet™ Butterfly Needle with Adapter 3/4 12 in.
Catalog No. 22025727
Change view
Click to view available options
Color:
Blue
Cream
Green
Orange
Needle Gauge:
19 G
21 G
23 G
25 G
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Needle Gauge
22-025-727 Cream 19 G
22-025-724 Green 21 G
22-025-725 Blue 23 G
22-025-726 Orange 25 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-025-727 Supplier Sol-Millennium Medical Supplier No. 110301070004
Add to Cart
Add to Cart
The butterfly needle features a removable multi-sample luer adapter, making its application flexible across multiple uses.
  • Thin-walled needle minimizes patient discomfort
  • Transparent design allows for clear visualization of blood flow
  • Available as a safety engineered (Sol-Care Safety Blood Collection Needle) device with a sliding safety shield that clicks when activated, telling you the needle is safely locked
  • Available with or without pre-attached blood collection tube holder
Specifications
Product Type Vet Butterfly
Color Cream
Material Plastic, Stainless Steel
Needle Gauge 19 G
Sterility Sterile
Latex Free Yes