Learn More
ATCC Sol8; Myoblast; Mouse (Mus musculus)
Catalog No. 502384925
Encompass
Sol8 are myoblasts isolated from the skeletal muscle of a 4-week-old mouse. It has applications in researching skeletal and cardiac muscle function.
Catalog No. 50-238-4925 Supplier ATCC Supplier No. CRL2174
 Restricted Item
Add to Cart
Add to Cart
Sol8 are myoblasts isolated from the skeletal muscle of a 4-week-old mouse. It has applications in researching skeletal and cardiac muscle function.