Learn More
Thermo Scientific™ SOLA™ SPE Cartridges
Catalog No. 03254075 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
SOLA HRP SPE Cartridge
SOLA HRP SPE Cartridge 30 mg/3 mL
SOLA SAX SPE Cartridge
SOLA SAX SPE Cartridge 30 mg/3 mL
SOLA SCX SPE Cartridge
SOLA SCX SPE Cartridge 30 mg/3 mL
SOLA WAX SPE Cartridge 30 mg/3 mL
SOLA WCX SPE Cartridge
SOLA WCX SPE Cartridge 30 mg/3 mL
Sorbent:
HRP (Reversed Phase Polymeric)
Mixed Mode Strong Anion Exchanger, Polymeric
Mixed Mode Strong Cation Exchanger, Polymeric
Mixed Mode Weak Anion Exchanger, Polymeric
Mixed Mode Weak Cation Exchanger, Polymeric
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Sorbent
03-254-075 SOLA SAX SPE Cartridge 30 mg/3 mL Mixed Mode Strong Anion Exchanger, Polymeric
03-150-391 SOLA HRP SPE Cartridge HRP (Reversed Phase Polymeric)
03-254-073 SOLA HRP SPE Cartridge 30 mg/3 mL HRP (Reversed Phase Polymeric)
03-150-392 SOLA SCX SPE Cartridge Mixed Mode Strong Cation Exchanger, Polymeric
03-254-074 SOLA SCX SPE Cartridge 30 mg/3 mL Mixed Mode Strong Cation Exchanger, Polymeric
03-150-393 SOLA SAX SPE Cartridge Mixed Mode Strong Anion Exchanger, Polymeric
60-109-004 SOLA WCX SPE Cartridge Mixed Mode Weak Cation Exchanger, Polymeric
03-254-099 SOLA WCX SPE Cartridge 30 mg/3 mL Mixed Mode Weak Cation Exchanger, Polymeric
03-254-100 SOLA WAX SPE Cartridge 30 mg/3 mL Mixed Mode Weak Anion Exchanger, Polymeric
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 03-254-075 Supplier Thermo Scientific™ Supplier No. 60409003
Add to Cart
Add to Cart
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Grow your laboratory productivity. Thermo Scientific™ SOLA™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
More about SOLA SPE cartridges
SOLA products are available in a 2mg/1mL, 10mg/1mL and 10mg/3mL cartridge formats. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of SOLA cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
SOLA cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What SOLA Cartridges Mean to Your Analysis
  • Prevent sample failures
  • Prevent blocked wells or cartridges
  • Reduced reanalysis
TRUSTED_SUSTAINABILITY
Specifications
Description SOLA SAX SPE Cartridge 30 mg/3 mL
Bed Weight 30 mg
For Use With (Application) Bioanalytical Applications - Strong Ion-Exchange Retention of Weak Acidic Compounds. Complementary Reversed Phase Retention of Neutral Compounds.
Product Line SOLA
Sorbent Mixed Mode Strong Anion Exchanger, Polymeric
Type SPE Cartridge
Volume (Metric) 3 mL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.