Learn More
Medchemexpress LLC HY-19436 5mg Medchemexpress, Solabegron CAS:252920-94-8 Purity:>98%
SDP
Catalog No. 501970025
Encompass_Preferred
Medchemexpress, HY-19436 5mg Solabegron CAS:252920-94-8 Solabegron (GW 427353) is a selective β3-adrenergic receptor agonist, stimulating cAMP accumulation in Chinese hamster ovary cells expressing the human β3-AR, with an EC50 value of 22 nM. Solabegron (GW 427353) is being developed for the treatment of overactive bladder and irritable bowel syndrome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0025 Supplier Medchemexpress LLC Supplier No. HY194365MG
Medchemexpress, HY-19436 5mg Solabegron CAS:252920-94-8 Solabegron (GW 427353) is a selective β3-adrenergic receptor agonist, stimulating cAMP accumulation in Chinese hamster ovary cells expressing the human β3-AR, with an EC50 value of 22 nM. Solabegron (GW 427353) is being developed for the treatment of overactive bladder and irritable bowel syndrome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.